Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Cytomegalovirus IE 70 kDa Antigen Monoclonal Antibody (D64G)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA17023
Description
MA1-7023 detects the Cytomegalovirus. This antibody is specific for a 70 kDa Immediate Early (IE) nuclear antigen. MA1-7023 has been successfully used in immunofluorescence procedures. The MA1-7023 immunogen is Cytomegalovirus lysate.
Cytomegalovirus (CMV) is an opportunistic pathogen infecting lung, kidney, gut and other organs in situations where an individual is immunologically immature, such as the foetus and neonate. Infection also occurs in immunosuppressed patients e.g. transplant patients, patients undergoing chemotherapy and those with HIV infection. Cytomegalovirus is an enveloped double stranded DNA virus which in humans (HCMV) is commonly known as human herpesvirus 5 (HHV-5).
Specifications
Cytomegalovirus IE 70 kDa Antigen | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG1 |
Immunocytochemistry | |
D64G | |
Unconjugated | |
CMV | |
Cytomegalovirus lysate. | |
1 mL | |
Primary | |
4°C | |
Liquid |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction