Learn More
Specifications
Specifications
| Concentration | 1 mg/ml |
| For Use With (Application) | ELISA, SDS-PAGE |
| Formulation | 0.2 % Triton X-100, 10mM Tris-HCl, 1mM EDTA and 100mM NaCl, pH 7.8 |
| Quantity | 0.5 mg |
| Storage Requirements | Store at -80C. Avoid freeze-thaw cycles. |
| Gene Alias | CMV; HHV 5; HHV5; Human herpesvirus 5 |
| Research Category | Virology Bacteria and Parasites |
| Protein | Cytomegalovirus |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
