Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytomegalovirus pp65 Monoclonal Antibody (1-L-11)

Catalog No. 01670331
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-670-331 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-670-331 Supplier Invitrogen™ Supplier No. MA17597
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-7597 detects HCMV from virus samples. MA1-7597 has been successfully used in ELISA, Western blot procedures. The MA1-7597 immunogen is HCMV infected cells. Store short-term at 4ºC, long-term at -80ºC.

Cytomegalovirus (CMV) is an opportunistic pathogen infecting lung, kidney, gut and other organs in situations where an individual is immunologically immature, such as the foetus and neonate. Infection also occurs in immunosuppressed patients e.g. transplant patients, patients undergoing chemotherapy and those with HIV infection. Cytomegalovirus is an enveloped double stranded DNA virus which in humans (HCMV) is commonly known as human herpesvirus 5 (HHV-5).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytomegalovirus pp65
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 1-L-11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.01% sodium azide; pH 7.2
Gene Alias CMV; Cytomegalovirus
Host Species Mouse
Immunogen HCMV infected cells.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C short term, -80°C long term
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.