Learn More
CytoScan™ MTT Cell Proliferation Assay, 1000 reactions, G-Biosciences
SCI_ED
Catalog No. S001116
Change view
Click to view available options
Quantity:
1000 Reactions
Catalog No. Quantity
S001116 1000 Reactions
1 options
Catalog No. S001116 Supplier G-Biosciences Supplier No. 7862257
Add to Cart
Add to Cart
CytoScan™ MTT Cell Proliferation Assay is a sensitive and accurate assay for cell proliferation and cytotoxicity. The assay is highly convenient as it is performed in a single cell tissue culture well and requires no washing, harvesting or solubilization of cells.
  • Adherent or suspension cells are cultured in a microplate and then incubated with MTT and the assay is monitored with a spectrophotometer. The assay principle is based upon the reduction of water-soluble MTT (3-(4,5-dimethylthiazol-2-yl)-2,5- diphenyltetrazolium bromide) to formazan by cellular dehydrogenases. Formazan is then solubilized, and the concentration of the colorimetric probe is determined by measuring optical density at 570 nm.
Specifications
For Use With (Application) Cell Proliferation
For Use With (Equipment) Spectrophotometer
Product Type MTT Cell Proliferation Assay
Quantity 1000 Reactions