Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Simport™ Scientific CytoSep™ Cytology Funnel for StatSpin™ Cytofuge Cytocentrifuge 2

Catalog No. 23045573
Click to view available options
:
Cytology Funnel for Cytocentrifuge 2; Type: Cytology Funnel; Filter Type: Paper; Filtration Area: 7 mm; For Use With (Equipment): StatSpin™ Cytofuge 2; Material (Membrane): Polypropylene
This item is not returnable. View return policy
This item is not returnable. View return policy

This cytology funnel allows cells to gently concentrate in a 7 mm diameter area on the slide, while the supernatant is simultaneously absorbed by a filter card.

  • Will accept 50 to 400 μL of sample
  • Produces cell mono layer presentations of high quality
  • Disposable funnels offer lower risk of contamination than reusable funnels
  • Disposable filters are sold separately

Specifications

Color Clear
Disposable Single-use
Filter Type Paper
For Use With (Equipment) StatSpin Cytofuge 2
Material (Membrane) Polypropylene
Sterility Non-sterile
Filtration Area 7 mm
For Use With (Application) Depositing Cells on Microscope Slide
Product Line CytoSep™
Type Cytology Funnel
Fit for Purpose Certified Quality
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.