Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytosolic beta-Glucosidase/GBA3 Antibody (019), DyLight 650 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289925C
Description
Cytosolic beta-Glucosidase/GBA3 Monoclonal antibody specifically detects Cytosolic beta-Glucosidase/GBA3 in Human samples. It is validated for ELISASpecifications
Cytosolic beta-Glucosidase/GBA3 | |
Monoclonal | |
DyLight 650 | |
CBG, CBGL1GLUC, cytosolic beta-glucosidase, Cytosolic beta-glucosidase-like protein 1, EC 3.2.1.21, glucosidase, beta, acid 3 (cytosolic), klotho-related protein, KLrP, MGC104276, MGC126878 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Cytosolic beta-Glucosidase/GBA3 (NP_066024.1; Met 1-Leu 469). | |
0.1 mL | |
metabolism | |
57733 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
019 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction