Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytosolic beta-Glucosidase/GBA3 Antibody (OTI1F1), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272100R
Description
Cytosolic beta-Glucosidase/GBA3 Monoclonal specifically detects Cytosolic beta-Glucosidase/GBA3 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
Cytosolic beta-Glucosidase/GBA3 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
GBA3 | |
Recombinant protein fragment corresponding to amino acids 1-150 and 370-469 of human Cytosolic beta-Glucosidase/GBA3 (NP_066024) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG2a |
Western Blot, Flow Cytometry, Immunohistochemistry | |
OTI1F1 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
CBG, CBGL1GLUC, cytosolic beta-glucosidase, Cytosolic beta-glucosidase-like protein 1, EC 3.2.1.21, glucosidase, beta, acid 3 (cytosolic), klotho-related protein, KLrP, MGC104276, MGC126878 | |
Mouse | |
Affinity Purified | |
metabolism | |
57733.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction