Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytosolic beta-Glucosidase/GBA3 Rabbit anti-Human, Clone: 8, Novus Biologicals™

Rabbit Monoclonal Antibody
$353.50 - $560.50
Specifications
Antigen | Cytosolic beta-Glucosidase/GBA3 |
---|---|
Clone | 8 |
Dilution | Flow Cytometry 1:25-1:100, Immunocytochemistry/Immunofluorescence 1:20-1:100 |
Applications | Flow Cytometry, Immunofluorescence |
Classification | Monoclonal |
Description
Cytosolic beta-Glucosidase/GBA3 Monoclonal specifically detects Cytosolic beta-Glucosidase/GBA3 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
Cytosolic beta-Glucosidase/GBA3 | |
Flow Cytometry 1:25-1:100, Immunocytochemistry/Immunofluorescence 1:20-1:100 | |
Monoclonal | |
Purified | |
RUO | |
0.2 um filtered solution in PBS | |
57733 | |
IgG | |
Protein A purified |
8 | |
Flow Cytometry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human | |
CBG, CBGL1GLUC, cytosolic beta-glucosidase, Cytosolic beta-glucosidase-like protein 1, EC 3.2.1.21, glucosidase, beta, acid 3 (cytosolic), klotho-related protein, KLrP, MGC104276, MGC126878 | |
Recombinant Human Cytosolic beta-Glucosidase/GBA3 protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title