Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytosolic Sulfotransferase 1A1/SULT1A1 Antibody (029), DyLight 755 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289949IR
Description
Cytosolic Sulfotransferase 1A1/SULT1A1 Monoclonal antibody specifically detects Cytosolic Sulfotransferase 1A1/SULT1A1 in Human samples. It is validated for ELISASpecifications
Cytosolic Sulfotransferase 1A1/SULT1A1 | |
Monoclonal | |
DyLight 755 | |
Aryl sulfotransferase 1, EC 2.8.2, EC 2.8.2.1, HAST1/HAST2, Phenol sulfotransferase 1, Phenol-sulfating phenol sulfotransferase 1, P-PST, P-PST 1, PST, ST1A1, ST1A3, STP1MGC5163, STPMGC131921, sulfotransferase 1A1, sulfotransferase family, cytosolic, 1A, phenol-preferring, member 1, Thermostable phenol sulfotransferase, thermostable phenol sulfotransferase1, ts-PST, TSPST1 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Cytosolic Sulfotransferase 1A1/SULT1A1 (P50225-1; Glu 2-Leu 295). | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA | |
029 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
6817 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction