Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytosolic Sulfotransferase 1A1/SULT1A1 Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299855G
Description
Cytosolic Sulfotransferase 1A1/SULT1A1 Polyclonal specifically detects Cytosolic Sulfotransferase 1A1/SULT1A1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Cytosolic Sulfotransferase 1A1/SULT1A1 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
6817 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 488 | |
Aryl sulfotransferase 1, EC 2.8.2, EC 2.8.2.1, HAST1/HAST2, Phenol sulfotransferase 1, Phenol-sulfating phenol sulfotransferase 1, P-PST, P-PST 1, PST, ST1A1, ST1A3, STP1MGC5163, STPMGC131921, sulfotransferase 1A1, sulfotransferase family, cytosolic, 1A, phenol-preferring, member 1, Thermostable phenol sulfotransferase, thermostable phenol sulfotransferase1, ts-PST, TSPST1 | |
Produced in rabbits immunized with purified, recombinant Cytosolic Sulfotransferase 1A1/SULT1A1 (P50225-1; Glu 2-Leu 295). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction