Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytosolic Sulfotransferase 1A1/SULT1A1 Antibody (OTI9B7), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271308PE
Description
Cytosolic Sulfotransferase 1A1/SULT1A1 Monoclonal specifically detects Cytosolic Sulfotransferase 1A1/SULT1A1 in Human samples. It is validated for Flow Cytometry.Specifications
Cytosolic Sulfotransferase 1A1/SULT1A1 | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
SULT1A1 | |
Full length recombinant protein of human Cytosolic Sulfotransferase 1A1/SULT1A1 (NP_001046) produced in HEK293T cells. | |
0.1 mL | |
6817.0 | |
Store at 4°C in the dark. |
Flow Cytometry | |
OTI9B7 | |
Flow Cytometry | |
Aryl sulfotransferase 1, EC 2.8.2, EC 2.8.2.1, HAST1/HAST2, Phenol sulfotransferase 1, Phenol-sulfating phenol sulfotransferase 1, P-PST, P-PST 1, PST, ST1A1, ST1A3, STP1MGC5163, STPMGC131921, sulfotransferase 1A1, sulfotransferase family, cytosolic, 1A, phenol-preferring, member 1, Thermostable phenol sulfotransferase, thermostable phenol sulfotransferase1, ts-PST, TSPST1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction