Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytosolic Sulfotransferase 1B1/SULT1B1 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299836PCP
Description
Cytosolic Sulfotransferase 1B1/SULT1B1 Polyclonal antibody specifically detects Cytosolic Sulfotransferase 1B1/SULT1B1 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
Cytosolic Sulfotransferase 1B1/SULT1B1 | |
Polyclonal | |
ELISA, Immunohistochemistry-Paraffin | |
EC 2.8.2, EC 2.8.2.-, EC 2.8.2.1, EC 2.8.2.4, MGC13356, ST1B1, ST1B2sulfotransferase family cytosolic 1B member 1, Sulfotransferase 1B1, Sulfotransferase 1B2, sulfotransferase family, cytosolic, 1B, member 1, SULT1B2, Thyroid hormone sulfotransferase | |
Produced in rabbits immunized with purified, recombinant Human Cytosolic Sulfotransferase 1B1/SULT1B1 (Accession#: NP_055280.2; Leu 2-Ile 296) | |
0.1 mL | |
metabolism | |
27284 | |
Store at 4C in the dark. | |
IgG |
ELISA, Immunohistochemistry (Paraffin) | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction