Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytosolic Sulfotransferase 1C2/SULT1C2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP153102
Description
Cytosolic Sulfotransferase 1C2/SULT1C2 Polyclonal specifically detects Cytosolic Sulfotransferase 1C2/SULT1C2 in Human samples. It is validated for Western Blot.Specifications
Cytosolic Sulfotransferase 1C2/SULT1C2 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
cytosolic, 1C, member 1, EC 2.8.2, EC 2.8.2.-, EC 2.8.2.1, EC 2.8.2.2, humSULTC2, ST1C2, Sulfotransferase 1C1, sulfotransferase family, cytosolic, 1C, member 2, SULT1C#1 | |
Rabbit | |
35 kDa | |
100 μL | |
metabolism | |
6819 | |
Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
O00338 | |
SULT1C2 | |
Synthetic peptides corresponding to SULT1C2(sulfotransferase family, cytosolic, 1C, member 2) The peptide sequence was selected from the C terminal of SULT1C2 (NP_001047). Peptide sequence LDQSISSFMRKGTVGDWKNHFTVAQNERFDEIYRRKMEGTSINFCMEL. | |
Affinity purified | |
RUO | |
Primary | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction