Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytosolic Sulfotransferase 1E1/SULT1E1 Antibody (011), Janelia Fluor™ 549, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289869JF549
Description
Cytosolic Sulfotransferase 1E1/SULT1E1 Monoclonal specifically detects Cytosolic Sulfotransferase 1E1/SULT1E1 in Human samples. It is validated for Sandwich ELISA Detection.Specifications
Cytosolic Sulfotransferase 1E1/SULT1E1 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
11 | |
Sandwich ELISA Detection | |
EC 2.8.2, EST-1, ESTMGC34459, estrone sulfotransferase, ST1E1, STEestrogen sulfotransferase, Sulfotransferase 1E1, sulfotransferase family 1E, estrogen-preferring, member 1, Sulfotransferase, estrogen-preferringEC 2.8.2.4 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Cytosolic Sulfotransferase 1E1/SULT1E1 (NP_005411.1; Asn2-Ile294). | |
0.1 mL | |
Breast Cancer, Stem Cell Markers | |
6783 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction