Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytosolic Sulfotransferase 1E1/SULT1E1 Antibody (CPTC-SULT1E1-1), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308212IR
Description
Cytosolic Sulfotransferase 1E1/SULT1E1 Monoclonal antibody specifically detects Cytosolic Sulfotransferase 1E1/SULT1E1 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
Cytosolic Sulfotransferase 1E1/SULT1E1 | |
Monoclonal | |
DyLight 755 | |
EC 2.8.2, EST-1, ESTMGC34459, estrone sulfotransferase, ST1E1, STEestrogen sulfotransferase, Sulfotransferase 1E1, sulfotransferase family 1E, estrogen-preferring, member 1, Sulfotransferase, estrogen-preferringEC 2.8.2.4 | |
Recombinant full-length human Cytosolic Sulfotransferase 1E1/SULT1E1 protein (Uniprot: P49888) | |
100 μg | |
Breast Cancer, Stem Cell Markers | |
6783 | |
Store at 4°C in the dark. | |
IgG2b |
Western Blot, Immunohistochemistry (Paraffin) | |
CPTC-SULT1E1-1 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction