Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Cytosolic Sulfotransferase 1E1/SULT1E1 Antibody Pair [HRP]
SDP

Catalog No. NB009667 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
15 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB009667 15 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB009667 Supplier Novus Biologicals™ Supplier No. NBP27931415PLATES
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Cytosolic Sulfotransferase 1E1/SULT1E1.

Specifications

Assay Range 31.25 pg/mL
Assay Sensitivity 31.25-2000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Cytosolic Sulfotransferase 1E1/SULT1E1.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias EC 2.8.2, EST-1, ESTMGC34459, estrone sulfotransferase, ST1E1, STEestrogen sulfotransferase, Sulfotransferase 1E1, sulfotransferase family 1E, estrogen-preferring, member 1, Sulfotransferase, estrogen-preferringEC 2.8.2.4
Gene ID (Entrez) 6783
Gene Symbol SULT1E1
Host Species Rabbit
Kit Contents
  • Standard: Each vial contains 56 ng of recombinant Cytosolic Sulfotransferase 1E1/SULT1E1.
  • Capture Antibody: 0.2 mg/mL of rabbit anti-Cytosolic Sulfotransferase 1E1/SULT1E1 monoclonal antibody,
  • Detection Antibody: 0.5 mg/mL rabbit anti-Cytosolic Sulfotransferase 1E1/SULT1E1 monoclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Breast Cancer, Stem Cell Markers
    Storage Requirements Storage is content dependent.
    Target Cytosolic Sulfotransferase 1E1/SULT1E1
    Technique 1. Capture Antibody: Dilute to a working concentration of 0.5 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.25 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 2 ng/mL is recommended.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.