Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytosolic Sulfotransferase 2A1/SULT2A1 Antibody (AT13E10), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP261163G
This item is not returnable.
View return policy
Description
Cytosolic Sulfotransferase 2A1/SULT2A1 Monoclonal antibody specifically detects Cytosolic Sulfotransferase 2A1/SULT2A1 in Human samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
Cytosolic Sulfotransferase 2A1/SULT2A1 | |
Monoclonal | |
DyLight 488 | |
alcohol/hydroxysteroid sulfotransferase, Dehydroepiandrosterone sulfotransferase, DHEAS, DHEA-STbile salt sulfotransferase, EC 2.8.2.14, HST, hSTa, Hydroxysteroid Sulfotransferase, ST2, ST2A1, ST2A3, STDbile-salt sulfotranasferase 2A1, Sulfotransferase 2A1, sulfotransferase family, cytosolic, 2A, dehydroepiandrosterone(DHEA)-preferring, member 1 | |
Recombinant human Cytosolic Sulfotransferase 2A1/SULT2A1 (1-285aa) purified from E.coli. | |
0.1 mL | |
Cancer, Endocrinology, Signal Transduction | |
6822 | |
Store at 4°C in the dark. | |
IgG2a κ |
Western Blot, ELISA, Immunofluorescence | |
AT13E10 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction