Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytosolic Sulfotransferase 2A1/SULT2A1 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299792H
Description
Cytosolic Sulfotransferase 2A1/SULT2A1 Polyclonal specifically detects Cytosolic Sulfotransferase 2A1/SULT2A1 in Human samples. It is validated for Western Blot, ELISA.Specifications
Cytosolic Sulfotransferase 2A1/SULT2A1 | |
Polyclonal | |
Western Blot, ELISA | |
alcohol/hydroxysteroid sulfotransferase, Dehydroepiandrosterone sulfotransferase, DHEAS, DHEA-STbile salt sulfotransferase, EC 2.8.2.14, HST, hSTa, Hydroxysteroid Sulfotransferase, ST2, ST2A1, ST2A3, STDbile-salt sulfotranasferase 2A1, Sulfotransferase 2A1, sulfotransferase family, cytosolic, 2A, dehydroepiandrosterone(DHEA)-preferring, member 1 | |
Produced in rabbits immunized with purified, recombinant Cytosolic Sulfotransferase 2A1/SULT2A1 (NP_003158.2; Ser 2-Glu 285). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, ELISA | |
HRP | |
PBS | |
Rabbit | |
Affinity purified | |
RUO | |
6822 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction