Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytosolic Sulfotransferase 2A1/SULT2A1 Antibody (OTI4D7), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271326IR
Description
Cytosolic Sulfotransferase 2A1/SULT2A1 Monoclonal specifically detects Cytosolic Sulfotransferase 2A1/SULT2A1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
Cytosolic Sulfotransferase 2A1/SULT2A1 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SULT2A1 | |
Full length recombinant protein of human Cytosolic Sulfotransferase 2A1/SULT2A1 (NP_003158) produced in HEK293T cells. | |
0.1 mL | |
6822.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
OTI4D7 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
alcohol/hydroxysteroid sulfotransferase, Dehydroepiandrosterone sulfotransferase, DHEAS, DHEA-STbile salt sulfotransferase, EC 2.8.2.14, HST, hSTa, Hydroxysteroid Sulfotransferase, ST2, ST2A1, ST2A3, STDbile-salt sulfotranasferase 2A1, Sulfotransferase 2A1, sulfotransferase family, cytosolic, 2A, dehydroepiandrosterone(DHEA)-preferring, member 1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction