Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Cytosporone B
GREENER_CHOICE

Catalog No. 545910
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
54-591-0 10 mg
54-595-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 54-591-0 Supplier Tocris Bioscience™ Supplier No. 5459/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

High affinity Nur77 agonist

Cytosporone B is a naturally occurring Nur77 (NR4A1) agonist (Kd = 8.52 x 10-7 M). Induces apoptosis in gastric cancer cells in vitro. Increases blood glucose levels in fasting C57 mice and suppresses xenograft tumor growth. Also exhibits a potent antifibrotic effect in bleomycin-induced skin fibrosis in vivo.

Specifications

Chemical Name or Material Ethyl 3,5-dihydroxy-2-(1-oxooctyl)-benzeneacetate
CAS 321661-62-5
Quantity 10 mg
Target Nuclear Receptor Agonists
Molecular Formula C18H26O5
Formula Weight 322.4
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.