Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab CytoStat Red Fixative
SDP

Catalog No. STLCSR1
Click to view available options
Quantity:
1 gal.
120 mL Prefill
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

An alcohol-based general purpose cytology fixative solution.

  • CytoStat Red is used for diagnostic non-gynecological cytology specimens, which include, but are not limited to, sputum, washings, body fluids, scrapings, and fine needle aspiration biopsies
  • This fixative lyses red blood cells and solubilizes proteins while preserving the diagnostically relevant cells
  • Gallon, 4 per case

Specifications

For Use With (Equipment) All Tissue Processors
Grade Chemical Solution
Product Type Fixative
Quantity 1 gal.
Solution Type Fixative
For Use With (Application) Fixation
Form Liquid
Format Solution
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.