Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
StatLab CytoStat Red Fixative

Click to view available options
Quantity:
1 gal.
120 mL Prefill
Description
- CytoStat Red is used for diagnostic non-gynecological cytology specimens, which include, but are not limited to, sputum, washings, body fluids, scrapings, and fine needle aspiration biopsies
- This fixative lyses red blood cells and solubilizes proteins while preserving the diagnostically relevant cells
- Gallon, 4 per case
Specifications
Specifications
For Use With (Equipment) | All Tissue Processors |
Grade | Chemical Solution |
Product Type | Fixative |
Quantity | 120 mL Prefill |
Solution Type | Fixative |
For Use With (Application) | Fixation |
Form | Liquid |
Format | Solution |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction