Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CytoVista™ Coronal Mouse Brain Slicer, 1 mm

Catalog No. V11335
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 slicer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V11335 1 slicer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. V11335 Supplier Invitrogen™ Supplier No. V11335
Add to Cart
Add to Cart

This 1 mm coronal mouse brain slicer allows you to easily cut a mouse brain into smaller pieces for easier imaging with objectives that have a limited working distance.

This slicer is 3D-printed out of resin and can be easily washed and cleaned between uses. All you need is a standard razor blade to start slicing your tissues.

Features of the CytoVista family of tissue and 3D cell culture clearing reagents include:
• Rapid clearing of fluorescently labeled tissue for 3D imaging
• Minimal shrinkage, expansion, or other morphological changes to tissue
• Simple workflow does not require any special instruments or equipment and is easily adapted to IHC
• Compatible with most fluorophores, including dyes and fluorescent proteins

The CytoVista tissue and 3D cell culture clearing workflow clears tissue or 3D cultured cells with minimal changes to morphology or the sensitivity of detection with almost any fluorophore. With an easy-to-use protocol, whole mouse brain (8-mm section) can be cleared in 24 hours, or a 1-mm section in 2 hours, without the use of any special instruments or equipment.

The CytoVista clearing workflow is compatible with most fluorophores, including fluorescent proteins that are detected with common fluorescent imaging instruments, such as wide-field microscopes, high content analyzers, confocal microscopes, and lightsheet microscopes. For first time users, we recommend the CytoVista Tissue Clearing/Staining Kit (Cat No V11324) or CytoVista 3D Cell Culture Clearing/Staining Kit (Cat No. V11325). These kits contain all necessary reagents for both clearing and IHC staining.

CytoVista clearing technology sufficiently clears tissue for 3D fluorescent imaging, but unlike other clearing techniques does not significantly changes the tissue morphology. Minimal morphological changes, such as shrinkage and contraction, have been observed. For precious samples, the clearing can be reversed, and the tissue can be processed for histology studies, like H&E staining.

For tissues that are easier to clear, like brain, lung, intestine, skin, and tissues of less than 250-μm thickness, treatment with CytoVista Tissue Clearing Reagent is sufficient for 3D imaging. However, tissues of more than 250-μm thickness or that are hard to clear like kidney, liver, heart, and placenta will require an initial treatment with CytoVista Tissue Clearing Reagent followed by treatment with CytoVista Tissue Clearing Enhancer.

CytoVista 3D Cell Culture Clearing Reagent is recommended for spheroids or organoids up to 1 mm in thickness.

For hard tissues like bone, cartilage, and connective tissue, CytoVista Tissue Clearing Reagent is not recommended.

TRUSTED_SUSTAINABILITY

Specifications

Type Coronal Mouse Brain Slicer
Product Line Image-iT
Content And Storage Store at room temperature.
Quantity 1 slicer
Shipping Condition Room Temperature
Can I autoclave the CytoVista Coronal Mouse Brain Slicer, 1 mm?

No, the CytoVista Coronal Mouse Brain Slicer, 1 mm cannot be autoclaved.

How can I clean the CytoVista Coronal Mouse Brain Slicer, 1 mm?

Clean with soap/detergent and cold water, rinse thoroughly and dry completely after cleaning. Expose to water only long enough to clean the slicer; do not soak in water for prolonged periods. Sonication and heat may distort the slicer.

What type of material is the CytoVista Coronal Mouse Brain Slicer, 1 mm made of?

It is composed of PLA (polylactic acid).

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.