Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ CyTRAK Orange™
SDP

Catalog No. NB016661 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB016661 200 μL
NB016662 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB016661 Supplier Novus Biologicals™ Supplier No. NBP281127200UL
Add to Cart
Add to Cart

Specifications

Quantity 200 μL
Content And Storage Store at 4°C in the dark. Do not freeze.
For Use With (Application) Flow Cytometry, Immunocytochemistry/Immunofluorescence, Live Imaging Microscopy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.