Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eBioscience™ CyTRAK Orange™

Catalog No. 5016969
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-169-69 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-169-69 Supplier Invitrogen™ Supplier No. 65088192
  • $320.00
    $239.65 / Each of 1

    Save $80.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Anthraquinone dye with high affinity for double-stranded DNA. It is a membrane-permeable dye that can label live or fixed/dead cells.

  • In flow cytometry, it can be used to distinguish nucleated and non-nucleated cells
  • In fluorescent microscopy, it can be used to identify and discriminate the nucleus and cytoplasm without the need for a second dye due to its high intensity staining of the nucleus and low intensity staining of the cytoplasm
  • CyTRAK Orange™ is optimally excited from 488 to 550nm and has a peak emission of 610 nm
  • It can be excited with the blue laser (488nm)
  • For a filter configuration, CyTRAK Orange™ is best detected with a 610/20 band pass filter or equivalent.
TRUSTED_SUSTAINABILITY

Specifications

Color Orange
Detection Method Fluorescence
For Use With (Application) Immunohistochemistry (IHC), Flow Cytometry
For Use With (Equipment) Fluorescence Microscope
Product Type Dye
Product Line eBioscience
Quantity 50 μL
Emission Wavelength 488 to 550/610

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.