Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYYR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA580536
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA580536 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA580536 Supplier Invitrogen™ Supplier No. PA580536
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody has specificity for Human CYYR1.

CYYR1 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include integral component of membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYYR1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene CYYR1
Gene Accession No. Q96J86
Gene Alias C21orf95; cysteine and tyrosine rich 1; cysteine and tyrosine-rich 1; cysteine and tyrosine-rich 1 protein; cysteine and tyrosine-rich protein 1; cysteine and tyrosine-rich protein 1 isoform 1,2,2bis,3,4; cysteine and tyrosine-rich protein 1 isoform 1,2,3,4b; cysteine and tyrosine-rich protein 1 isoform 1,2,3b; cysteine and tyrosine-rich protein 1 isoform 1,2,4; cysteine/tyrosine-rich 1; Cyyr1; hypothetical protein zgc:77252; LOW QUALITY PROTEIN: cysteine and tyrosine-rich protein 1; proline-rich domain-containing protein; zgc:77252
Gene Symbols CYYR1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the C-terminus of the Human CYYR1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116159
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.