Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYYR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564336
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564336 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564336 Supplier Invitrogen™ Supplier No. PA564336
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MKNHRATRVG ILRTTHINTV SSYPAGPPPY GHDHEMEYCA DLPPPYSPTP QGPAQR Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 68%.

CYYR1 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include integral component of membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYYR1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CYYR1
Gene Accession No. Q96J86
Gene Alias C21orf95; cysteine and tyrosine rich 1; cysteine and tyrosine-rich 1; cysteine and tyrosine-rich 1 protein; cysteine and tyrosine-rich protein 1; cysteine and tyrosine-rich protein 1 isoform 1,2,2bis,3,4; cysteine and tyrosine-rich protein 1 isoform 1,2,3,4b; cysteine and tyrosine-rich protein 1 isoform 1,2,3b; cysteine and tyrosine-rich protein 1 isoform 1,2,4; cysteine/tyrosine-rich 1; Cyyr1; hypothetical protein zgc:77252; LOW QUALITY PROTEIN: cysteine and tyrosine-rich protein 1; proline-rich domain-containing protein; zgc:77252
Gene Symbols CYYR1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CYYR1. Recombinant protein control fragment (Product #RP-103815).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116159
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.