Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CZC 25146
GREENER_CHOICE

Catalog No. 607110
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-711-0 10 mg
60-715-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 60-711-0 Supplier Tocris Bioscience™ Supplier No. 6071/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent LRRK2 inhibitor

CZC 25146 is a potent LRRK2 inhibitor (IC50 values are 4.76 and 6.87 nM for for wild-type and G2019S mutant forms of LRRK2, respectively). Also inhibits PLK4, GAK, TNK1, CAMKK2 and PIP4K2. Attenuates mutant LRRK2-induced injury of cultured rodent and human neurons. Cell permeable.

Specifications

Chemical Name or Material CZC 25146
CAS 1191911-26-8
Synonym N-[2-[[5-Fluoro-2-[[2-methoxy-4-(4-morpholinyl)phenyl]amino]-4-pyrimidinyl]amino]phenyl]methanesulfonamide
Quantity 10 mg
Solubility Soluble to 100mM in DMSO
Molecular Formula C22H25FN6O4S
Formula Weight 488.54
Physical Form Solid
Color Pale-purple
Product Type Bioactive Small Molecule
Purity >98%
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.