Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-2-Aminoadipic acid, 98%, Thermo Scientific™

Catalog No. p-4323513
Click to view available options
Quantity:
5 g

Chemical Identifiers

CAS 7620-28-2
Molecular Formula C6H11NO4
Molecular Weight (g/mol) 161.16
InChI Key OYIFNHCXNCRBQI-SCSAIBSYSA-N
Synonym d-2-aminoadipic acid, r-2-aminohexanedioic acid, 2r-2-aminohexanedioic acid, d-a-aminoadipic acid, d-, a-aminoadipic acid, r-2-aminoadipic acid, r-2-amino-hexanedioic acid, hexanedioic acid, 2-amino-,--and +, hexanedioic acid, 2-amino-, 2r, d-homoglutamic acid
PubChem CID 165627
ChEBI CHEBI:37025
IUPAC Name (2R)-2-aminohexanedioic acid
SMILES C(CC(C(=O)O)N)CC(=O)O

Specifications

Melting Point 186°C to 188°C
Color White
Elemental Analysis 43.60 to 45.84% C, 8.47 to 8.91% N
Infrared Spectrum Authentic
Assay Percent Range 98%
Quantity 5 g
Specific Rotation -20.5
Formula Weight 161.16
Percent Purity 98%
Physical Form Crystalline Powder or Powder
Chemical Name or Material D-2-Aminoadipic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.