Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D.Aspartic acid Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988624
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8624 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8624 Supplier Bioss Supplier No. BS8529R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

D-Aspartic acid is the D-isomer of aspartic acid also known as aspartate which is one of the 20 natural proteinogenic amino acids, the building blocks of proteins. As its name indicates, aspartic acid is the carboxylic acid analog of asparagine. It is non-essential in mammals, and might serve as an excitatory neurotransmitter in the brain. It is also a metabolite in the urea cycle, and participates in gluconeogenesis.

Specifications

Antigen D.Aspartic acid
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Host Species Rabbit
Immunogen D.Aspartic acid conjugated to KLH.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.