Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Biotin, MP Biomedicals

Catalog No. p-7112280
Click to view available options
Quantity:
100 mg
500 mg

  • D-Biotin may be used to elute proteins from avidin/streptavidin resins
  • It is widely used for dietary supplements and fortified foods
  • It is also used for tablets and hard-shell capsule preparation due to its pharmaceutical properties

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-85-5
Molecular Formula C10H16N2O3S
Molecular Weight (g/mol) 244.31
MDL Number MFCD00005541
InChI Key YBJHBAHKTGYVGT-UHFFFAOYNA-N
Synonym biotin, d-biotin, vitamin h, coenzyme r, vitamin b7, bios ii, factor s, bioepiderm, d +-biotin, biodermatin
PubChem CID 171548
ChEBI CHEBI:15956
IUPAC Name 5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanoic acid
SMILES OC(=O)CCCCC1SCC2NC(=O)NC12

Specifications

Melting Point 223°C to 233°C
Color White
Boiling Point 573.6°C (760 mmHg(literature))
Linear Formula C10H16N2O3S
Quantity 100 mg
Solubility Information Soluble in water (22 mg/100 mL), ethanol (80 mg/100 mL), more soluble in hot water and in dilute alkalies; insoluble in other common organic solvents. Soluble in 2 M Ammonium hydroxide (50mg/mL - clear, colorless solution), dimethylformamide (1.7mg/mL).
Formula Weight 244.3
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material D-BIOTIN
Recommended Storage Store at +4°C.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.