Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
D and C Red No. 33, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
25 g
100 g
500 g
5 kg
Packaging:
Amber Glass Bottle
Description
Spectrum™ Chemical D and C Red No. 33
Specifications
Specifications
CAS Max % | 100% |
Grade | Reagent |
Packaging | Amber Glass Bottle |
Color | Red |
Quantity | 500 g |
Chemical Name or Material | D and C Red No. 33 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction