Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D and C Red No. 6, 90%, Spectrum™ Chemical
SDP

Catalog No. 18601986 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-986 25 g Amber Glass Bottle
18-601-987 100 g Amber Glass Bottle
18-601-988 500 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-601-986 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. D113925GM
Add to Cart
Add to Cart

D1139, 5858-81-1, C18H14N2O6S·2Na

Spectrum™ Chemical D and C Red No. 6 is used as a food dye and originally used as a replacement for amaranth as a food coloring. It looks like a dark red powder and often comes as a sodium salt. In the United States, the Food and Drug Administration (FDA) controls the specifications for Drug and Cosmetic colors. Spectrum certifies all batches of artificial drug and cosmetic colors meet FDA D&C quality.

Chemical Identifiers

CAS 5858-81-1
Molecular Formula C18H12N2Na2O6S
Molecular Weight (g/mol) 430.34
InChI Key CTBASIXHDUUSCV-SDXCCTRSSA-L
IUPAC Name disodium (4E)-4-[2-(4-methyl-2-sulfonatophenyl)hydrazin-1-ylidene]-3-oxo-3,4-dihydronaphthalene-2-carboxylate
SMILES [Na+].[Na+].CC1=CC=C(N\N=C2\C(=O)C(=CC3=CC=CC=C23)C([O-])=O)C(=C1)S([O-])(=O)=O

Specifications

CAS Max % 100%
Percent Purity 90%
Grade Reagent
Packaging Amber Glass Bottle
Color Red
Quantity 25 g
Chemical Name or Material D and C Red No. 6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.