Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
D and C Red No. 6, 90%, Spectrum™ Chemical

Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Amber Glass Bottle
Description
Spectrum™ Chemical D and C Red No. 6 is used as a food dye and originally used as a replacement for amaranth as a food coloring. It looks like a dark red powder and often comes as a sodium salt. In the United States, the Food and Drug Administration (FDA) controls the specifications for Drug and Cosmetic colors. Spectrum certifies all batches of artificial drug and cosmetic colors meet FDA D&C quality.
Specifications
Specifications
CAS Max % | 100% |
Percent Purity | 90% |
Grade | Reagent |
Packaging | Amber Glass Bottle |
Color | Red |
Quantity | 100 g |
Chemical Name or Material | D and C Red No. 6 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction