Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D(+)-Camphor, 97%

Catalog No. p-82156
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 464-49-3 | C10H16O | 152.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 464-49-3
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.24
MDL Number MFCD00064149,MFCD00074738,MFCD00064149,MFCD00064149
InChI Key DSSYKIVIOFKYAU-XVKPBYJWSA-N
Synonym +-camphor, d-camphor, 1r-+-camphor, r-camphor, +-bornan-2-one, r-+-camphor, 1r,4r-1,7,7-trimethylbicyclo 2.2.1 heptan-2-one, 1r,4r-camphor, camphor d, +-2-bornanone
PubChem CID 159055
ChEBI CHEBI:15396
IUPAC Name (1R,4R)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-one
SMILES CC1(C)[C@H]2CC[C@@]1(C)C(=O)C2

Specifications

Melting Point 176.0°C to 180.0°C
Color White
Boiling Point 204.0°C
Flash Point 64°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Quantity 100 g
Fieser 13,61
Merck Index 15, 1734
Specific Rotation + 42.50
Solubility Information Solubility in water: 1g/800mL water (25°C) (giving a colloidal solutio. Other solubilities: at 25°C 1 g dissolves in 1 mL alcohol,1 mL ether,,0.5 mL chloroform,0.4 mL benzene,0.4 mL acetone,,1.5 mL turpentine oil,0.5 mL glacial acetic acid
Formula Weight 152.24
Percent Purity 97%
Physical Form Crystals
Chemical Name or Material D(+)-Camphor, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable solid.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cauti
EINECSNumber 207-355-2
RTECSNumber EX1260000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.