Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-(+)-Celloheptaose, 95%, Thermo Scientific Chemicals

Catalog No. AAJ67803LB0
Change view
Click to view available options
Quantity:
1 mg
2 mg
5 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67803LB0 5 mg
AAJ67803MCR 1 mg
AAJ678038PL 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ67803LB0 Supplier Thermo Scientific Chemicals Supplier No. J67803LB0

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52646-27-2
MDL Number MFCD15145329
Synonym Celloheptaose

Specifications

Color White
Quantity 5 mg
Sensitivity Hygroscopic
Formula Weight 1153.02
Percent Purity 95%
Physical Form Powder
Chemical Name or Material D-(+)-Celloheptaose
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.