Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Cysteine hydrochloride monohydrate, 98%

Catalog No. p-4630204
Click to view available options
Quantity:
250 mg
500 mg
1 g
Packaging:
Glass Bottle

CAS: 207121-46-8 | C3H10ClNO3S | 175.627 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207121-46-8
Molecular Formula C3H10ClNO3S
Molecular Weight (g/mol) 175.627
MDL Number MFCD00150051
InChI Key QIJRTFXNRTXDIP-YBBRRFGFSA-N
Synonym d-cysteine hydrochloride monohydrate, s-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, d-cysteine*hcl*h2o, d-cysteine hydrochloride hydrate, h-d-cys-oh.hcl.h2o, d-cysteine hydrate hydrochloride, c3h7no2s.hcl.h2o, d-cysteine hcl monohydrate, pubchem12314, h-d-cys-oh??hcl??h2o
PubChem CID 10176341
IUPAC Name (2S)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride
SMILES C(C(C(=O)O)N)S.O.Cl

Specifications

Color White to Yellow
Packaging Glass Bottle
Quantity 250 mg
Formula Weight 177.77
Physical Form Crystalline Powder
Assay 98%
Chemical Name or Material D-Cysteine hydrochloride monohydrate
EINECSNumber 251-043-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.