Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
D-Dimer Antibody (28), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP105045AF594
Description
D-Dimer Monoclonal specifically detects D-Dimer in Human, Canine samples. It is validated for Western Blot, ELISA.Specifications
D-Dimer | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
Mouse | |
0.1 mL | |
Binds human D-dimer. Cross-reacts with directly coated D-monomer but not with directly coated fibrinogen. It also reacts with dog D-dimer. | |
Store at 4°C in the dark. |
Western Blot, ELISA | |
28 | |
Western Blot, ELISA | |
D Dimer | |
Purified human D-dimer adsorbed onto aluminum hydroxide gel. | |
Primary | |
Human, Canine | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction