Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
D-Dimer Antibody (DD2), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1108376AF594
Description
D-Dimer Monoclonal antibody specifically detects D-Dimer in Human samples. It is validated for Western Blot, ELISA, Immunoassay, ELISASpecifications
D-Dimer | |
Monoclonal | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Human | |
Purified |
Western Blot, ELISA, Immunoassay, ELISA | |
DD2 | |
Western Blot, ELISA, Immunoassay, Sandwich ELISA | |
D Dimer | |
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with D-dimer. | |
0.2 mL | |
Primary | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction