Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
D-Dimer Antibody (DD41cc), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10073038H
Description
D-Dimer Monoclonal specifically detects D-Dimer in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA.Specifications
D-Dimer | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
Mouse | |
Protein A purified | |
RUO | |
D-dimer and high molecular weight fibrin degradation products, no cross-reactivity with fibrinogen | |
Store at 4C in the dark. | |
IgG2a |
Western Blot, ELISA | |
DD41cc | |
Western Blot, ELISA, Sandwich ELISA | |
D Dimer | |
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with mixture of D-dimer and high molecular weight fibrin degradation products. | |
0.2 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction