Learn More
Description
Specifications
Specifications
| Antigen | D-Dimer |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | DD41cc |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100-1:2000, Sandwich ELISA 1:100-1:2000 |
| Gene Alias | D Dimer |
| Host Species | Mouse |
| Immunogen | Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with mixture of D-dimer and high molecular weight fibrin degradation products. |
| Purification Method | Protein A purified |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
