Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ D-dimer Chimeric Recombinant Rabbit Monoclonal Antibody (3B6)
GREENER_CHOICE

Catalog No. PIMA547781
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547781 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547781 Supplier Invitrogen™ Supplier No. MA547781
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Specificity: Recognises epitopes that reside between amino acids 86 and 302 in the gamma-chain of fragment D chains of fibrinogen (Fragment D) and other cross-linked fibrin degradation products containing D-Dimer, but no reactivity with Fragment D or E, fibrinogen or fibrinogen degradation products.

Fibrinogen is the main protein of blood coagulation system. It consists of two identical subunits that contain three polypeptide chains: alpha, beta and gamma. The process of blood coagulation results in the activation of fibrinogen into fibrin by thrombin and fibrin polymerization. Fibrin clot is then digested by plasmin, and fibrin degradation products of different molecular weights are released into the bloodstream.
TRUSTED_SUSTAINABILITY

Specifications

Antigen D-dimer Chimeric
Applications ELISA, Lateral Flow
Classification Recombinant Monoclonal
Clone 3B6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene FGA
Gene Accession No. P02671
Gene Alias carboxy-terminal region; ENSMUSG00000059807; FGA; Fib; Fib2; fibrinogen A-alpha-chain; fibrinogen alpha chain; fibrinogen, A alpha polypeptide; Fibrinopeptide A; MGC119422; MGC119423; MGC119425
Gene Symbols FGA
Host Species Rabbit
Immunogen Human cross-linked D-Dimer.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2243
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.