Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ D-dimer Monoclonal Antibody (DD6)
GREENER_CHOICE

Catalog No. PIMA120031
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA120031 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA120031 Supplier Invitrogen™ Supplier No. MA120031
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-20031 detects D-Dimer in human samples.

Fibrinogen is the main protein of blood coagulation system. It consists of two identical subunits that contain three polypeptide chains: alpha, beta and gamma. The process of blood coagulation results in the activation of fibrinogen into fibrin by thrombin and fibrin polymerization. Fibrin clot is then digested by plasmin, and fibrin degradation products of different molecular weights are released into the bloodstream.
TRUSTED_SUSTAINABILITY

Specifications

Antigen D-dimer
Applications ELISA, Western Blot
Classification Monoclonal
Clone DD6
Concentration 4.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene FGA
Gene Accession No. E9PV24
Gene Alias carboxy-terminal region; ENSMUSG00000059807; FGA; Fib; Fib2; fibrinogen A-alpha-chain; fibrinogen alpha chain; fibrinogen, A alpha polypeptide; Fibrinopeptide A; MGC119422; MGC119423; MGC119425
Gene Symbols FGA
Host Species Mouse
Immunogen High molecular weight fibrin degradation products or synthetic peptides covering the cross-linked region of D-dimer gamma-chain.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14161
Target Species Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.