Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Toluidine, 99%, crystalline molten mass

Catalog No. AC376350250
Click to view available options
Quantity:
250 g
1 kg
5 kg
25 kg
Packaging:
Glass Bottle
HDPE Bottle
Metal drum

CAS: 106-49-0 | C7H9N | 107.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-49-0
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.16
InChI Key RZXMPPFPUUCRFN-UHFFFAOYSA-N
Synonym p-toluidine, 4-toluidine, p-tolylamine, 4-aminotoluene, benzenamine, 4-methyl, 4-methylbenzenamine, p-methylaniline, p-methylbenzenamine, p-toluidin, p-aminotoluene
PubChem CID 7813
ChEBI CHEBI:37825
IUPAC Name 4-methylaniline
SMILES CC1=CC=C(C=C1)N

Specifications

Melting Point 41.0°C to 46.0°C
Color White to Brown
Boiling Point 200.0°C
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Metal drum
Linear Formula CH3C6H4NH2
Quantity 25 kg
Merck Index 15,9696
Solubility Information Solubility in water: 1.1 g/100mL. Other solubilities: freely soluble in diluted acids, freely soluble in alcohol, ether, acetone, carbon, disulfide, methanol and oils, 156% in ethanol
Formula Weight 107.16
Percent Purity 99%
Physical Form Crystalline Mass or Crystals
Assay 99%
Chemical Name or Material p-Toluidine, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic in contact with skin.
Suspected of causing cancer.
Causes serious eye irritation.
May cause an allergic skin reaction.
Toxic if inhaled.
Very toxic to aquatic life.
Toxic if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:
EINECSNumber 203-403-1
Recommended Storage Product may darken on storage

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.