Learn More
SHOWA™ 910 Stainless Steel/HPPE Cut-Resistant Gloves
Catalog No. 19048577A Shop All Showa Best Glove Products
Change view
Click to view available options
Size:
10
6
7
8
9
Medium
Coating Material:
PVC Dots
Uncoated
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Coating Material
19-048-577A 6 PVC Dots
19-048-577C 8 PVC Dots
19-048-577D 9 PVC Dots
19-048-577E 10 PVC Dots
19-048-576A 6 Uncoated
19-048-576D 9 Uncoated
19-048-576E 10 Uncoated
19-048-577B 7 PVC Dots
19-048-576C Medium Uncoated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 19-048-577A Supplier SHOWA™ Supplier No. 910C06
This item is not returnable. View return policy
Flexible and lightweight gloves are available in uncoated or PVC coated styles and are ideal for light-duty applications.
  • High-performance polyethylene engineered fiber for added strength, comfort and cut resistance
  • Fully launderable and color coded for easy identification
  • Gauge: 10
  • Uncoated or PVC dotted style available
Specifications
Certifications/Compliance (Europe) CE
Construction Seamless Knit
Coating Material PVC Dots
Color White
Cut Resistance Rating ANSI A4
Hand Ambidextrous
Material Polyethylene
Product Type Cut-resistant Gloves
Size 6
EN388 Standard EN 388:2X43DP
For Use With (Application) Food processing, machine or compartment assembly, bottling, sheet Metal handling, construction, recycling, Glass handling, razor blades
Glove Cuff Style Knit
Yarn Gauge 10
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.