Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D(+)-Galactosamine Hydrochloride, 99%, Thermo Scientific Chemicals

Catalog No. AC160442500
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle

D(+)-Galactosamine hydrochloride ((2R,3R,4R,5R)-2-amino-3,4,5,6-tetrahydroxyhexanal hydrochloride), CAS # 1772-03-8, is the salt of the amino sugar that acts as an RNA synthesis inhibitor in hepatocytes.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• D-Galactosamine is a pyranose amino sugar with hepatotoxic traits

• It acts as an RNA synthesis inhibitor in hepatocytes

• It promotes adherence of polymorphonuclear leukocytes to endothelial cells, increasing superoxide production

Applications

• D-Galactosamine can be used as a hepatotoxic in experimental in vitro models when in combination with LPS

• It is suitable for a range of biochemical laboratory applications

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1772-03-8
Molecular Formula C6H13NO5·HCl
MDL Number MFCD00135830
Synonym Chondrosamine hydrochloride, 2-Amino-2-deoxy-D-galactose hydrochloride

Specifications

Color White
Ignition Residue 1% max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 250 mg
Specific Rotation Condition +96° (20°C 589nm c=1,H2O,24 hr.)
Specific Rotation +93° to +99° (20°C, 589nm) (c=1, H2O), after 24 hrs.
Formula Weight 215.64
Percent Purity ≥98.5%
Physical Form Fine Crystalline Powder
Chemical Name or Material D(+)-Galactosamine hydrochloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.