Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
D(+)-Galactose, 99+%
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General description
- Galactose is a monosaccharide and aldohexose found in most organisms
- Galactose is an epimer of glucose and can be converted to glucose through the Leloir pathway
- It also plays a role as a metabolite in mice and E. coli
- Galactose is less stable than glucose and rapid conversion often occurs
- Galactose may also be incorporated into glycoconjugates
Applications
- Metabolism of galactose may be relevant to the study of aging, inflammation, and metabolic disorders
- It is intended for a variety of applications from in vitro metabolic assays to HPLC
Specifications
Specifications
| Melting Point | 165.0°C to 168.0°C |
| Color | White |
| Density | 6.2 |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99+% |
| Quantity | 100 g |
| Beilstein | 31, 295 |
| Merck Index | 15, 4365 |
| Specific Rotation Condition | + 79.75 (20.00°C c=10,H2O,NH3) |
| Specific Rotation | + 79.75 |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction