Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D(+)-Galactose, 99+%, Acros Organics™

Click to view available options
Quantity:
100g
500g
1kg
5kg
25kg
59-23-4
C6H12O6
MFCD00063833
59-23-4
C6H12O6
MFCD00063833
Infrared Spectrum Authentic
Specific Rotation + 79.75
Melting Point 165.0°C to 168.0°C
Appearance (10% aq. soln.) clear, colorless
Water 0.3% max. (K.F.)
Quantity 500g
Sulfated Ash 0.1% max. (on dry substance)
Specific Rotation Condition + 79.75 (20.00°C c=10,H2O,NH3)
Vapor Pressure Negligible
Color White
Density 6.2
Assay Percent Range 99+%
Beilstein 31, 295
Merck Index 15, 4365
Formula Weight 180.16
Physical Form Powder
Percent Purity ≥99%
Chemical Name or Material D(+)-Galactose
Show More Show Less