Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Galactose, Anhydrous, Reagent, 99%, Spectrum™ Chemical
SDP

Catalog No. 18602124 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
100 g
500 g
1 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle

GA110, 59-23-4, C6H12O6

Spectrum™ Chemical D-Galactose, Anhydrous, Reagent

Chemical Identifiers

CAS 59-23-4
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
InChI Key WQZGKKKJIJFFOK-UHFFFAOYNA-N
IUPAC Name 6-(hydroxymethyl)oxane-2,3,4,5-tetrol
SMILES OCC1OC(O)C(O)C(O)C1O

Specifications

CAS Max % 100%
Packaging Poly Bottle
Quantity 1 kg
Percent Purity 99%
Grade Reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.