Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α-D-Glucoheptonic Acid γ-lactone 99%, Thermo Scientific™

Catalog No. AC154931000
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC154931000 100 g
AC154935000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC154931000 Supplier Thermo Scientific Chemicals Supplier No. 154931000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60046-25-5
Molecular Formula C7H12O7
Molecular Weight (g/mol) 208.166
InChI Key VIVCRCODGMFTFY-VGHHYEQRSA-N
Synonym d-glucoheptono-1,4-lactone, d-glucoheptonolactone, glucoheptonic acid, a-lactone, 2, i-d-glucoheptonic acid, 3s,5s-3,4-dihydroxy-5-1r,2r-1,2,3-trihydroxypropyl oxolan-2-one
PubChem CID 71317081
IUPAC Name (3S,5S)-3,4-dihydroxy-5-[(1R,2R)-1,2,3-trihydroxypropyl]oxolan-2-one
SMILES C(C(C(C1C(C(C(=O)O1)O)O)O)O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 152°C to 155°C
Color White
Infrared Spectrum Authentic
Quantity 100 g
Formula Weight 208.17
Percent Purity ≥98.5%
Physical Form Fine Crystalline Powder
Chemical Name or Material α-D-Glucoheptonic acid γ-lactone, 99%
EINECSNumber 262-037-0
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.